Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

1-Phenylimidazole ≥99%

Supplier: MATRIX SCIENTIFIC MS

004505-1G 004505-5G
101787-344EA 52.31 USD InStock
101787-344 101787-346
1-Phenylimidazole ≥99%
1-Phenylimidazole
Hazard code(s): Irritant


Formula: C₉H₈N₂
Boiling Pt: 142 °C (15mm)
Melting Pt: 13 °C
Density: 1.1397 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00041204
CAS Number: 7164-98-9
UN: 2384
ADR: 3,II
Order Now
Specification Test Results
Boiling point 142°C/15mm
Melting point 13°C
Density 1.12

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty