Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2,4-Dibromo-6-(trifluoromethyl)aniline ≥97%

Supplier: MATRIX SCIENTIFIC MS

004560-5G 004560-25G
101787-486EA 40 USD InStock
101787-486 101787-488
2,4-Dibromo-6-(trifluoromethyl)aniline ≥97%
2,4-Dibromo-6-(trifluoromethyl)aniline
Hazard code(s): Irritant


Formula: C₇H₄Br₂F₃N
Boiling Pt: 58 °C (0.7mm)
MDL Number: MFCD00178769
CAS Number: 71757-14-7
UN: 2811
ADR: 6.1,III
Order Now
Specification Test Results
Boiling point 58°C/0.7mm

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty