Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

(3-Iodophenyl)acetonitrile ≥97% for synthesis

Supplier: MATRIX SCIENTIFIC MS

004718-1G 004718-5G
101787-732EA 107.69 USD InStock
101787-732 101787-734
(3-Iodophenyl)acetonitrile ≥97% for synthesis
(3-Iodophenyl)acetonitrile
Hazard code(s): Irritant, Irritant-Harmful, Light Sensitive


Formula: C₈H₆IN
MW: 243.05
Boiling Pt: 138
Melting Pt: 34…36 °C
MDL Number: MFCD00040890
CAS Number: 130723-54-5
UN: 3439
ADR: 6.1 III
Order Now
Specification Test Results
Boiling point 138°C/12mm
Melting point 34-36°C

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty