Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

4-Vinylbenzoic acid ≥92%

Supplier: MATRIX SCIENTIFIC MS
Warning

004733-1G 004733-5G 004733-25G
101787-756EA 26.15 USD InStock
101787-756 101787-758 101787-760
4-Vinylbenzoic acid ≥92%
4-Vinylbenzoic acid
Hazard code(s): Irritant, Store in Freezer


Formula: C₉H₈O₂
Boiling Pt: 282 °C (1013 hPa)
Melting Pt: 115 °C
Density: 1.3 g/cm³ (20 °C)
Storage Temperature: Refrigerator
MDL Number: MFCD00002569
CAS Number: 1075-49-6
EINECS: 214-053-4
Order Now
Specification Test Results
Melting point 143-144°C

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty