Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2,2,3,3-Tetrachlorohexafluorobutane ≥99%

Supplier: MATRIX SCIENTIFIC MS
Warning

004774-5G 004774-25G
101787-790EA 52.31 USD InStock
101787-790 101787-792
2,2,3,3-Tetrachlorohexafluorobutane ≥99%
2,2,3,3-Tetrachlorohexafluorobutane
Hazard code(s): Irritant


Formula: C₄Cl₄F₆
Boiling Pt: 131…133 °C
Melting Pt: 83…84 °C
Density: 1.76 g/cm³
MDL Number: MFCD00042088
CAS Number: 375-34-8
EINECS: 206-787-9
Order Now
Specification Test Results
Boiling point 131-133°C
Melting point 83-84°C
Density 1.76

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty