Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

1-Iodoperfluorooctane ≥98%

Supplier: MATRIX SCIENTIFIC MS
Warning

004859-25G 004859-100G
101787-912EA 38.46 USD InStock
101787-912 101787-914
1-Iodoperfluorooctane ≥98%
1-Iodoperfluorooctane
Hazard code(s): Irritant, Light Sensitive, Refrigerate


Formula: C₈F₁₇I
Boiling Pt: 160 °C (1013 hPa)
Melting Pt: 25 °C
Density: 2.071 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00001064
CAS Number: 507-63-1
EINECS: 208-079-5
Order Now
Specification Test Results
Boiling point 160-161°C

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty