Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Hexafluoroglutaryl dichloride ≥98%

Supplier: MATRIX SCIENTIFIC MS

004880-1G 004880-5G
101787-948EA 115.38 USD InStock
101787-948 101787-950
Hexafluoroglutaryl dichloride ≥98%
Hexafluoroglutaryl dichloride
Hazard code(s): Corrosive


Formula: C₅Cl₂F₆O₂
Boiling Pt: 103 - 107 °C (1013 hPa)
Density: 1.6389 g/cm³ (20 °C)
MDL Number: MFCD00039309
CAS Number: 678-77-3
EINECS: 211-651-7
UN: 3265
ADR: 8,II
Order Now
Specification Test Results
Boiling point 111°C
Density 1.64
ND20 1.354

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty