Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2,4-Dichloro-3,5-dinitro-α,α,α-trifluorotoluene ≥98% for synthesis

Supplier: MATRIX SCIENTIFIC MS

005005-1G 005005-5G
101788-146EA 38.46 USD InStock
101788-146 101788-148
2,4-Dichloro-3,5-dinitro-α,α,α-trifluorotoluene ≥98% for synthesis
2,4-Dichloro-3,5-dinitro-α,α,α-trifluorotoluene
Hazard code(s): Irritant-Harmful


Formula: C₇HCl₂F₃N₂O₄
Boiling Pt: 291 - 294 °C (1013 hPa)
Melting Pt: 74 - 75 °C
MDL Number: MFCD00042480
CAS Number: 29091-09-6
EINECS: 249-420-8
UN: 3077
ADR: 9,III
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty