Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2,4-Dichloro-3-nitrobenzotrifluoride ≥98% for synthesis

Supplier: MATRIX SCIENTIFIC MS

005023-1G 005023-5G
101788-178EA 72.31 USD InStock
101788-178 101788-180
2,4-Dichloro-3-nitrobenzotrifluoride ≥98% for synthesis
2,4-Dichloro-3-nitrobenzotrifluoride
Hazard code(s): Irritant, Irritant-Harmful


Formula: C₇H₂Cl₂F₃NO₂
MW: 260
MDL Number: MFCD00153114
CAS Number: 203915-49-5
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty