Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2,4-Dichloro-5-nitrobenzotrifluoride ≥97% for synthesis

Supplier: MATRIX SCIENTIFIC MS
Warning

005118-5G 005118-25G 005118-100G
101788-276EA 556.92 USD InStock
101788-276 101788-278 101788-280
2,4-Dichloro-5-nitrobenzotrifluoride ≥97% for synthesis
2,4-Dichloro-5-nitrobenzotrifluoride
Hazard code(s): Irritant, Irritant-Harmful


Formula: C₇H₂Cl₂F₃NO₂
MW: 260 g/mol
Melting Pt: 55…57 °C
MDL Number: MFCD00007073
CAS Number: 400-70-4
Order Now
Specification Test Results
Melting point 55-57°C

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty