Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

3,5-Dichlorobenzaldehyde ≥97% for synthesis

Supplier: MATRIX SCIENTIFIC MS
Warning

005161-5G 005161-25G 005161-100G
101788-340EA 18.46 USD InStock
101788-340 101788-342 101788-344
3,5-Dichlorobenzaldehyde ≥97% for synthesis
3,5-Dichlorobenzaldehyde
Hazard code(s): Irritant, Air Sensitive


Formula: C₇H₄Cl₂O
Boiling Pt: 215 - 222 °C (7 -rr)
Melting Pt: 63 °C
Storage Temperature: Ambient
MDL Number: MFCD00003352
CAS Number: 10203-08-4
EINECS: 233-499-0
Order Now
Specification Test Results
Melting point 64-66°C

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty