Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2,4-Difluorobenzenesulfonamide

Supplier: MATRIX SCIENTIFIC MS

005180-5G 005180-25G
101788-398EA 75.38 USD InStock
101788-398 101788-400
2,4-Difluorobenzenesulfonamide
2,4-Difluorobenzenesulphonamide


Formula: C₆H₅F₂NO₂S
MDL Number: MFCD01320743
CAS Number: 13656-60-5
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty