Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2,6-Difluorobenzophenone ≥98%

Supplier: MATRIX SCIENTIFIC MS
Warning

005215-5G 005215-25G
101788-484EA 43.08 USD InStock
101788-484 101788-486
2,6-Difluorobenzophenone ≥98%
2,6-Difluorobenzophenone
Hazard code(s): Irritant


Formula: C₁₃H₈F₂O
Boiling Pt: 120 °C (2mm)
Density: 1.24 g/cm³ (20 °C)
MDL Number: MFCD00009895
CAS Number: 59189-51-4
EINECS: 261-650-0
Order Now
Specification Test Results
Boiling point 120°C/2mm
Density 1.244
ND20 1.5645

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty