Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2,3-Difluorobenzoyl chloride ≥97%

Supplier: MATRIX SCIENTIFIC MS

005227-5G 005227-25G
101788-506EA 104.62 USD InStock
101788-506 101788-508
2,3-Difluorobenzoyl chloride ≥97%
2,3-Difluorobenzoyl chloride
Hazard code(s): Corrosive


Formula: C₇H₃ClF₂O
Boiling Pt: 85 - 87 °C (1013 hPa)
Density: 1.423 g/cm³ (20 °C)
MDL Number: MFCD00042499
CAS Number: 18355-73-2
UN: 3265
ADR: 8,II
Order Now
Specification Test Results
Boiling point 190°C
Density 1.423
ND20 1.514

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty