Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

3,5-Difluorobenzyl alcohol ≥97% for synthesis

Supplier: MATRIX SCIENTIFIC MS
Warning

005246-5G 005246-25G 005246-100G
101788-538EA 15.38 USD InStock
101788-538 101788-540 101788-542
3,5-Difluorobenzyl alcohol ≥97% for synthesis
3,5-Difluorobenzyl alcohol
Hazard code(s): Irritant


Formula: C₇H₆F₂O
Boiling Pt: 98…99 °C (9mm)
Density: 1.27 g/cm³ (20 °C)
MDL Number: MFCD00010313
CAS Number: 79538-20-8
Order Now
Specification Test Results
Boiling point 98-99°C/9mm
Density 1.272
ND20 1.487

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty