Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

(3-(Trifluoromethoxy)phenyl)acetic acid ≥98% for synthesis

Supplier: MATRIX SCIENTIFIC MS
Warning

005309-5G 005309-25G
101788-646EA 50.77 USD InStock
101788-646 101788-648
(3-(Trifluoromethoxy)phenyl)acetic acid ≥98% for synthesis
(3-(Trifluoromethoxy)phenyl)acetic acid
Hazard code(s): Irritant


Formula: (C₆H₄(OCF₃))CH₂COOH
MW: 220.15
Melting Pt: 53…55 °C
MDL Number: MFCD00082480
CAS Number: 203302-97-0
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty