Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2-(Trifluoromethoxy)phenylboronic acid ≥97% for synthesis

Supplier: MATRIX SCIENTIFIC MS

005343-5G 005343-25G 005343-100G
101788-694EA 21.54 USD InStock
101788-694 101788-696 101788-698
2-(Trifluoromethoxy)phenylboronic acid ≥97% for synthesis
2-(Trifluoromethoxy)phenylboronic acid
Hazard code(s): Moisture Sensitive, Store in Freezer


Formula: C₇H₆BF₃O₃
Melting Pt: 118…120 °C
MDL Number: MFCD01320763
CAS Number: 175676-65-0
Order Now
Specification Test Results
Melting point 118-120°C

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty