Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

4-(Trifluoromethoxy)benzonitrile ≥98%

Supplier: MATRIX SCIENTIFIC MS

005350-25G 005350-100G
101788-720EA 53.85 USD InStock
101788-720 101788-722
4-(Trifluoromethoxy)benzonitrile ≥98%
4-(Trifluoromethoxy)benzonitrile
Hazard code(s): Irritant, Irritant-Harmful


Formula: C₈H₄F₃NO
Boiling Pt: 192 °C (1013 hPa)
Density: 1.284 g/cm³ (20 °C)
MDL Number: MFCD00039474
CAS Number: 332-25-2
EINECS: 206-363-3
UN: 3276
ADR: 6.1,III
Order Now
Specification Test Results
Boiling point 192-193°C
Density 1.285
ND20 1.452

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty