Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2-(Trifluoromethoxy)benzaldehyde ≥95% for synthesis

Supplier: MATRIX SCIENTIFIC MS
Warning

005366-5G 005366-25G
101788-758EA 36.92 USD InStock
101788-758 101788-760
2-(Trifluoromethoxy)benzaldehyde ≥95% for synthesis
2-(Trifluoromethoxy)benzaldehyde
Hazard code(s): Irritant, Air Sensitive


Formula: C₈H₅F₃O₂
Boiling Pt: 77 °C (20mm)
Density: 1.330 g/cm³
MDL Number: MFCD00042405
CAS Number: 94651-33-9
Order Now
Specification Test Results
Boiling point 77°C/20mm
Density 1.33

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty