Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

α-Chloro-4-(trifluoromethoxy)toluene ≥97% for synthesis

Supplier: MATRIX SCIENTIFIC MS

005368-1G 005368-5G
101788-762EA 21.54 USD InStock
101788-762 101788-764
α-Chloro-4-(trifluoromethoxy)toluene ≥97% for synthesis
α-Chloro-4-(trifluoromethoxy)toluene
Hazard code(s): Corrosive, Refrigerate


Formula: C₈H₆ClF₃O
MW: 210.58
Boiling Pt: 81…83 °C (15mm)
MDL Number: MFCD00052326
CAS Number: 65796-00-1
UN: 3265
ADR: 8 II
Order Now
Specification Test Results
Boiling point 81-83°C/15m

 

ORDER

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty