Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

4-(Trifluoromethyl)cinnamic acid ≥98% for synthesis

Supplier: MATRIX SCIENTIFIC MS
Warning

005398-25G 005398-100G
101788-846EA 192.31 USD InStock
101788-846 101788-848
4-(Trifluoromethyl)cinnamic acid ≥98% for synthesis
4-(Trifluoromethyl)cinnamic acid
Hazard code(s): Irritant


Formula: C₁₀H₇F₃O₂
Melting Pt: 230…231 °C
MDL Number: MFCD00002696
CAS Number: 2062-26-2
Order Now
Specification Test Results
Melting point 230-231°C

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty