Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2-(Trifluoromethyl)phenylhydrazinium chloride ≥95% for synthesis

Supplier: MATRIX SCIENTIFIC MS

005424-5G 005424-25G
101788-930EA 32.31 USD InStock
101788-930 101788-932
2-(Trifluoromethyl)phenylhydrazinium chloride ≥95% for synthesis
2-(Trifluoromethyl)phenylhydrazinium chloride
Hazard code(s): Irritant


Formula: C₇H₈ClF₃N₂
Melting Pt: 220 °C
MDL Number: MFCD00102619
CAS Number: 3107-34-4
Order Now
Specification Test Results
Melting point 220°C

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty