Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

α,α,α-Trifluoro-2-tolualdehyde ≥98% for synthesis

Supplier: MATRIX SCIENTIFIC MS
Warning

005494-25G 005494-100G
101789-054EA 27.69 USD InStock
101789-054 101789-056
α,α,α-Trifluoro-2-tolualdehyde ≥98% for synthesis
α,α,α-Trifluoro-2-tolualdehyde
Hazard code(s): Irritant


Formula: C₈H₅F₃O
Boiling Pt: 28 - 38 °C (0.4 -rr)
Melting Pt: -40 °C
Density: 1.322 g/cm³ (20 °C)
MDL Number: MFCD00003337
CAS Number: 447-61-0
EINECS: 207-185-9
Order Now
Specification Test Results
Boiling point 70-71°C/15m
Density 1.32
ND20 1.466

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty