Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

(2,6-Dichlorophenyl)acetic acid ≥99% for synthesis

Supplier: MATRIX SCIENTIFIC MS
Warning

005575-25G 005575-100G
101789-158EA 18.46 USD InStock
101789-158 101789-160
(2,6-Dichlorophenyl)acetic acid ≥99% for synthesis
(2,6-Dichlorophenyl)acetic acid
Hazard code(s): Irritant


Formula: (C₆H₃Cl₂)CH₂COOH
Boiling Pt: 316 °C (1013 hPa)
Melting Pt: 160 - 161 °C
Storage Temperature: Ambient
MDL Number: MFCD00004320
CAS Number: 6575-24-2
EINECS: 229-504-0
Order Now
Specification Test Results
Melting point 158-161°C

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty