Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

(3,5-Difluorophenyl)acetic acid ≥99%

Supplier: MATRIX SCIENTIFIC MS
Warning

005582-25G 005582-100G
101789-190EA 176.92 USD InStock
101789-190 101789-192
(3,5-Difluorophenyl)acetic acid ≥99%
(3,5-Difluorophenyl)acetic acid
Hazard code(s): Irritant


Formula: (C₆H₃F₂)CH₂COOH
Melting Pt: 68…70 °C
MDL Number: MFCD00010316
CAS Number: 105184-38-1
Order Now
Specification Test Results
Melting point 68-70°C

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty