Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2,6-Dichlorophenylboronic acid for synthesis

Supplier: MATRIX SCIENTIFIC MS
Warning

005593-5G 005593-25G
101789-210EA 32.31 USD InStock
101789-210 101789-212
2,6-Dichlorophenylboronic acid for synthesis
2,6-Dichlorophenylboronic acid


Formula: C₆H₅BCl₂O₂
MW: 190.82
MDL Number: MFCD00064869
CAS Number: 73852-17-2
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty