Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

3-Chlorophenethylamine ≥99%

Supplier: MATRIX SCIENTIFIC MS

005694-1G 005694-5G
101789-366EA 41.54 USD InStock
101789-366 101789-368
3-Chlorophenethylamine ≥99%
3-Chlorophenethylamine
Hazard code(s): Irritant, Irritant-Harmful


Formula: C₈H₁₀ClN
Boiling Pt: 92 °C (5mm)
Density: 1.12 g/cm³
MDL Number: MFCD00047957
CAS Number: 13078-79-0
EINECS: 235-982-1
Order Now
Specification Test Results
Boiling point 92°C/5mm
Density 1.12

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty