Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

3,5-Bis(trifluoromethyl)benzamide ≥97%

Supplier: MATRIX SCIENTIFIC MS

005819-5G 005819-25G
101789-592EA 55.38 USD InStock
101789-592 101789-594
3,5-Bis(trifluoromethyl)benzamide ≥97%
3,5-Bis(trifluoromethyl)benzamide
Hazard code(s): Irritant


Formula: C₉H₅F₆NO
Melting Pt: 160…162 °C
MDL Number: MFCD00015511
CAS Number: 22227-26-5
EINECS: 244-849-7
Order Now
Specification Test Results
Melting point 160-162°C

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty