Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

5-Iodo-α,α,α,α',α',α'-hexafluoro-m-xylene ≥97% for synthesis

Supplier: MATRIX SCIENTIFIC MS

005875-5G 005875-25G
101789-684EA 15.38 USD InStock
101789-684 101789-686
5-Iodo-α,α,α,α',α',α'-hexafluoro-m-xylene ≥97% for synthesis
5-Iodo-α,α,α,α',α',α'-hexafluoro-m-xylene
Hazard code(s): Irritant


Formula: C₈H₃F₆I
Boiling Pt: 171 - 172 °C (1013 hPa)
Density: 1.91 g/cm³ (20 °C)
MDL Number: MFCD00040837
CAS Number: 328-73-4
Order Now
Specification Test Results
Boiling point 62-64°C/12m

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty