Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

1-Bromo-3-(difluoromethoxy)benzene for synthesis

Supplier: MATRIX SCIENTIFIC MS

006010-5G 006010-25G
101789-910EA 60 USD InStock
101789-910 101789-912
1-Bromo-3-(difluoromethoxy)benzene for synthesis
1-Bromo-3-(difluoromethoxy)benzene


Formula: C₇H₅BrF₂O
MW: 223.02
MDL Number: MFCD00236233
CAS Number: 262587-05-3
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty