Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

3-Tolylboronic acid ≥97% for synthesis

Supplier: MATRIX SCIENTIFIC MS

006050-5G 006050-25G
101789-974EA 15.38 USD InStock
101789-974 101789-976
3-Tolylboronic acid ≥97% for synthesis
3-Tolylboronic acid
Hazard code(s): Irritant, Store in Freezer


Formula: C₇H₉BO₂
MW: 135.96 g/mol
Melting Pt: 165
MDL Number: MFCD00040198
CAS Number: 17933-03-8
Order Now
Specification Test Results
Melting point 69-71°C

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty