Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Tetrafluorophthalic acid ≥97%

Supplier: MATRIX SCIENTIFIC MS

006125-25G 006125-100G
101790-144EA 23.08 USD InStock
101790-144 101790-146
Tetrafluorophthalic acid ≥97%
Tetrafluorophthalic acid
Hazard code(s): Irritant


Formula: C₈H₂F₄O₄
Boiling Pt: 341 °C (1013 hPa)
Melting Pt: 162…164 °C
MDL Number: MFCD00002407
CAS Number: 652-03-9
EINECS: 211-483-4
Order Now
Specification Test Results
Melting point 162-164°C

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty