Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Trifluoro(1,1,2,2-tetrafluoro-2-bromoethoxy)ethylene ≥97%

Supplier: MATRIX SCIENTIFIC MS

006357-5G 006357-25G
101790-408EA 446.15 USD InStock
101790-408 101790-410
Trifluoro(1,1,2,2-tetrafluoro-2-bromoethoxy)ethylene ≥97%
Trifluoro(1,1,2,2-tetrafluoro-2-bromoethoxy)ethylene
Hazard code(s): Irritant, Refrigerate


Formula: F₂CCFOCF₂CF₂Br
Boiling Pt: 54…55 °C
MDL Number: MFCD02183517
CAS Number: 85737-06-0
EINECS: 288-516-4
UN: 3295
ADR: 3,III
Order Now
Specification Test Results
Boiling point 54-55°C

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty