Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

4'-Trifluoromethoxyacetophenone ≥98% for synthesis

Supplier: MATRIX SCIENTIFIC MS

006485-5G 006485-25G 006485-100G
101790-606EA 15.38 USD InStock
101790-606 101790-608 101790-610
4'-Trifluoromethoxyacetophenone ≥98% for synthesis
4'-Trifluoromethoxyacetophenone
Hazard code(s): Irritant


Formula: C₉H₇F₃O₂
MW: 204.15
Boiling Pt: 110
Density: 1.278 g/cm³
MDL Number: MFCD00042404
CAS Number: 85013-98-5
EINECS: 285-066-0
Order Now
Specification Test Results
Boiling point 47-49°C/15m
Density 1.278
ND20 1.455

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty