Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

4'-(Trifluoromethyl)acetophenone ≥98% for synthesis

Supplier: MATRIX SCIENTIFIC MS
Warning

006496-25G 006496-100G
101790-642EA 53.85 USD InStock
101790-642 101790-644
4'-(Trifluoromethyl)acetophenone ≥98% for synthesis
4'-(Trifluoromethyl)acetophenone
Hazard code(s): Irritant, Refrigerate


Formula: C₉H₇F₃O
MW: 188.15 g/mol
Boiling Pt: 205
Melting Pt: 30…33 °C
Density: 0.924 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00000401
CAS Number: 709-63-7
EINECS: 211-913-0
Order Now
Specification Test Results
Boiling point 79-80°C/8mm
Melting point 30-33°C
ND20 1.4604

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty