Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

1-Benzyloxy-4-iodobenzene ≥98% for synthesis

Supplier: MATRIX SCIENTIFIC MS

006556-5G 006556-25G
101790-734EA 141.54 USD InStock
101790-734 101790-736
1-Benzyloxy-4-iodobenzene ≥98% for synthesis
1-Benzyloxy-4-iodobenzene
Hazard code(s): Irritant


Formula: C₁₃H₁₁IO
MW: 310.13
Boiling Pt: 140 °C (1mm)
Melting Pt: 59…61 °C
MDL Number: MFCD00079704
CAS Number: 19578-68-8
Order Now
Specification Test Results
Boiling point 140°C/1mm
Melting point 59-61°C

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty