Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

3-Iodo-o-xylene ≥97% for synthesis

Supplier: MATRIX SCIENTIFIC MS
Warning

006560-5G 006560-25G
101790-750EA 69.23 USD InStock
101790-750 101790-752
3-Iodo-o-xylene ≥97% for synthesis
3-Iodo-o-xylene
Hazard code(s): Irritant


Formula: C₈H₉I
MW: 232.06 g/mol
Boiling Pt: 98…100 °C (15mm)
Density: 1.64 g/cm³
MDL Number: MFCD00041437
CAS Number: 31599-60-7
Order Now
Specification Test Results
Boiling point 98-100°C/15
Density 1.64
ND20 1.64

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty