Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

5-Fluoroisatin ≥97% for synthesis

Supplier: MATRIX SCIENTIFIC MS
Warning

007122-25G 007122-100G
101792-066EA 21.54 USD InStock
101792-066 101792-068
5-Fluoroisatin ≥97% for synthesis
5-Fluoroisatin
Hazard code(s): Irritant


Formula: C₈H₄FNO₂
MW: 165.12 g/mol
Melting Pt: 224…227 °C
Storage Temperature: Ambient
MDL Number: MFCD00022795
CAS Number: 443-69-6
Order Now
Specification Test Results
Melting point 224-227°C

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty