Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2,3-Bis(trifluoromethyl)pyridine for synthesis

Supplier: MATRIX SCIENTIFIC MS

007212-1G 007212-5G
101792-204EA 72.31 USD InStock
101792-204 101792-206
2,3-Bis(trifluoromethyl)pyridine for synthesis
2,3-Bis(trifluoromethyl)pyridine
Hazard code(s): Irritant


Formula: C₇H₃F₆N
Boiling Pt: 158 °C (1013 hPa)
Melting Pt: -2 °C
MDL Number: MFCD03093763
CAS Number: 1644-68-4
Order Now
Specification Test Results
Boiling point 85°C/18mm

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty