Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

3-(Trifluoromethyl)-1H-pyrazole ≥95% for synthesis

Supplier: MATRIX SCIENTIFIC MS

007256-5G 007256-25G
101792-264EA 20 USD InStock
101792-264 101792-266
3-(Trifluoromethyl)-1H-pyrazole ≥95% for synthesis
3-(Trifluoromethyl)-1H-pyrazole
Hazard code(s): Irritant


Formula: C₄H₃F₃N₂
MW: 136.08
Boiling Pt: 72 °C (120mm)
Melting Pt: 49…50 °C
MDL Number: MFCD00115018
CAS Number: 20154-03-4
Order Now
Specification Test Results
Boiling point 72°C/120mm
Melting point 49-50°C

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty