Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

5-Bromoindole ≥99%

Supplier: MATRIX SCIENTIFIC MS

007262-100G 007262-500G
101792-284EA 60 USD InStock
101792-284 102550-894
5-Bromoindole ≥99%
5-Bromoindole
Hazard code(s): Store in Freezer


Formula: C₈H₆BrN
Melting Pt: 90…92 °C
Storage Temperature: Ambient
MDL Number: MFCD00005670
CAS Number: 10075-50-0
EINECS: 233-208-7
Order Now
Specification Test Results
Melting point 90-92°C

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty