Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

3-Fluoroisonicotinic acid ≥98%

Supplier: MATRIX SCIENTIFIC MS

008694-1G 008694-5G 008694-25G
101795-190EA 15.38 USD InStock
101795-190 101795-192 101795-194
3-Fluoroisonicotinic acid ≥98%
3-Fluoroisonicotinic acid
Hazard code(s): Irritant


Formula: C₆H₄FNO₂
Melting Pt: 251…254 °C (dec.)
MDL Number: MFCD03789251
CAS Number: 393-53-3
Order Now
Specification Test Results
Melting point 251-254°C(d

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty