Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Methyl-4-iodobenzoate ≥98% for synthesis

Supplier: MATRIX SCIENTIFIC MS

008705-25G 008705-100G
101795-228EA 73.85 USD InStock
101795-228 101795-230
Methyl-4-iodobenzoate ≥98% for synthesis
Methyl-4-iodobenzoate
Hazard code(s): Irritant


Formula: C₈H₇IO₂
MW: 262.05
Boiling Pt: 276
Melting Pt: 112
Density: 2.02 g/cm³ (10 °C)
MDL Number: MFCD00016353
CAS Number: 619-44-3
EINECS: 210-597-1
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty