Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2-Pyridylmethanol ≥98% for synthesis

Supplier: MATRIX SCIENTIFIC MS
Warning

008725-25G 008725-100G
101795-296EA 21.54 USD InStock
101795-296 101795-298
2-Pyridylmethanol ≥98% for synthesis
2-Pyridylmethanol
Hazard code(s): Irritant, Refrigerate, Hygroscopic


Formula: C₆H₇NO
Boiling Pt: 112…113 °C (16mm)
Melting Pt: –6…–5 °C
Density: 1.1317 g/cm³ (20 °C)
MDL Number: MFCD00006348
CAS Number: 586-98-1
EINECS: 209-592-7
Order Now
Specification Test Results
Boiling point 112-113°C/1
Melting point -6 to -5°C
Density 1.131
ND20 1.544

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty