Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

6-Fluoroindole ≥98%

Supplier: MATRIX SCIENTIFIC MS
Warning

008771-5G 008771-25G
101795-416EA 18.46 USD InStock
101795-416 101795-418
6-Fluoroindole ≥98%
6-Fluoroindole
Hazard code(s): Irritant, Store in Freezer


Formula: C₈H₆FN
Melting Pt: 74…77 °C
Storage Temperature: Ambient
MDL Number: MFCD00056933
CAS Number: 399-51-9
Order Now
Specification Test Results
Melting point 74-77°C

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty