Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

4'-Chloro-3'-(trifluoromethyl)acetophenone ≥99%

Supplier: MATRIX SCIENTIFIC MS

008882-5G 008882-25G 008882-100G
101795-710EA 26.15 USD InStock
101795-710 101795-712 101795-714
4'-Chloro-3'-(trifluoromethyl)acetophenone ≥99%
4'-Chloro-3'-(trifluoromethyl)acetophenone
Hazard code(s): Irritant


Formula: C₉H₆ClF₃O
Melting Pt: 61…63 °C
MDL Number: MFCD03094146
CAS Number: 129825-11-2
Order Now
Specification Test Results
Melting point 61-63°C

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty