Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2-Chloro-3-fluorobenzonitrile ≥98% for synthesis

Supplier: MATRIX SCIENTIFIC MS

008951-5G 008951-25G 008951-100G
101795-900EA 21.54 USD InStock
101795-900 101795-902 101795-904
2-Chloro-3-fluorobenzonitrile ≥98% for synthesis
2-Chloro-3-fluorobenzonitrile
Hazard code(s): Irritant


Formula: C₇H₃ClFN
MW: 155.56
MDL Number: MFCD03407966
CAS Number: 874781-08-5
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty