Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Indole-4-carbonitrile for synthesis

Supplier: MATRIX SCIENTIFIC MS
Warning

009070-5G 009070-25G
101796-268EA 36.92 USD InStock
101796-268 101796-270
Indole-4-carbonitrile for synthesis
Indole-4-carbonitrile


Formula: C₉H₆N₂
MW: 142.16
MDL Number: MFCD00152045
CAS Number: 16136-52-0
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty