Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Perfluoro-1-octene ≥95% for synthesis

Supplier: MATRIX SCIENTIFIC MS

009123-1G 009123-5G
101796-412EA 223.08 USD InStock
101796-412 101796-414
Perfluoro-1-octene ≥95% for synthesis
Perfluoro-1-octene
Hazard code(s): Irritant


Formula: C₈F₁₆
MW: 400.06
Boiling Pt: 105 °C
MDL Number: MFCD00236708
CAS Number: 559-14-8
EINECS: 209-202-5
Order Now
Specification Test Results
Boiling point 105°C

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty