Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

1-Bromo-2-chloro-5-fluorobenzene ≥98% for synthesis

Supplier: MATRIX SCIENTIFIC MS
Warning

009434-5G 009434-25G 009434-100G
101797-102EA 23.08 USD InStock
101797-102 101797-104 101797-106
1-Bromo-2-chloro-5-fluorobenzene ≥98% for synthesis
1-Bromo-2-chloro-5-fluorobenzene
Hazard code(s): Irritant


Formula: C₆H₃BrClF
Density: 1.75 g/cm³
MDL Number: MFCD00061120
CAS Number: 201849-15-2
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty